LIVH93E9FC76 Liverpool ChiroChem

tert-butyl (R)-2-((methylamino)methyl)morpholine-4-carboxylate

  • Cas Number : 2277435-90-0
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A9878?search=A9878

Safety Information

MSDS https://www.liverpoolchirochem.com/A9878?search=A9878
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart