LIVH93E9FC78 Liverpool ChiroChem

2-(4-(piperidin-4-yl)phenyl)acetic acid hydrochloride

  • Cas Number : 34033-52-8
  • Molecular Weight : 255.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl salt
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A10529?search=A10529

Safety Information

MSDS https://www.liverpoolchirochem.com/A10529?search=A10529
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart