LIVH93E9FC7F Liverpool ChiroChem

tert-butyl 4-(4-(2-aminoethyl)phenyl)piperidine-1-carboxylate

  • Cas Number : 1402667-19-9
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/A9989?search=A9989

Safety Information

MSDS https://www.liverpoolchirochem.com/A9989?search=A9989
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart