LIVH93E9FC8E Liverpool ChiroChem

2-Fluoro-4-(methylsulfonyl)phenol

  • Cas Number : 398456-87-6
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/BC0223?search=BC0223

Safety Information

MSDS https://www.liverpoolchirochem.com/BC0223?search=BC0223
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart