LIVH93E9FC8F Liverpool ChiroChem

2-fluoro-1-methoxy-4-(methylsulfonyl)benzene

  • Cas Number : 20951-14-8
  • Molecular Weight : 204.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/BC0224?search=BC0224

Safety Information

MSDS https://www.liverpoolchirochem.com/BC0224?search=BC0224
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart