LIVH93E9FC99 Liverpool ChiroChem

Methyl 6-methoxypyrazine-2-carboxylate

  • Cas Number : 23813-24-3
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/KS0007?search=KS0007

Safety Information

MSDS https://www.liverpoolchirochem.com/KS0007?search=KS0007
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart