LIVH93E9FC9E Liverpool ChiroChem

1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde

  • Cas Number : 276862-85-2
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/LC0009?search=LC0009

Safety Information

MSDS https://www.liverpoolchirochem.com/LC0009?search=LC0009
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart