LIVH93E9FCA0 Liverpool ChiroChem

Methyl 1H-pyrrolo[3,2-b]pyridine-3-carboxylate

  • Cas Number : 952800-39-4
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/LC0012?search=LC0012

Safety Information

MSDS https://www.liverpoolchirochem.com/LC0012?search=LC0012
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart