LIVH93E9FCAB Liverpool ChiroChem

4-bromo-1-methyl-1H-1,2,3-triazole

  • Cas Number : 13273-53-5
  • Molecular Weight : 161.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.liverpoolchirochem.com/LC0073?search=LC0073

Safety Information

MSDS https://www.liverpoolchirochem.com/LC0073?search=LC0073
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart