MAN182106108 Manchester Organics

9-Nitro-1,2,3,4-tetrahydro-5H-1,4-benzodiazepin-5-one

  • MDL number MFCD04973407
  • Molecular Weight : 207.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J16209

Safety Information

MSDS https://www.manchesterorganics.com/J16209
Availability details for
Quantity
  • Availability
  • Add to Cart