MAN182106111 Manchester Organics

5-Pyridin-3-yl-4H-pyrazole-3-carboxylic acid

  • MDL number MFCD06245340
  • Molecular Weight : 189.17078

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S16122

Safety Information

MSDS https://www.manchesterorganics.com/S16122
Availability details for
Quantity
  • Availability
  • Add to Cart