MAN182106119 Manchester Organics

2,3-Diphenyl-quinoxaline-6-carboxylic acid

  • MDL number MFCD00555042
  • Molecular Weight : 326.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R16309

Safety Information

MSDS https://www.manchesterorganics.com/R16309
Availability details for
Quantity
  • Availability
  • Add to Cart