MAN182106135 Manchester Organics

Pyrazin-2-yl-thiourea

  • MDL number MFCD04971717
  • Molecular Weight : 154.193

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U16195
Availability details for
Quantity
  • Availability
  • Add to Cart