MAN182106179 Manchester Organics

5-(2-Methoxy-phenyl)-2H-pyrazole-3-carboxylic acid

  • MDL number MFCD03030186
  • Molecular Weight : 218.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P16546

Safety Information

MSDS https://www.manchesterorganics.com/P16546
Availability details for
Quantity
  • Availability
  • Add to Cart