MAN182106180 Manchester Organics

5-(3-Methoxy-phenyl)-2H-pyrazole-3-carboxylic acid methyl ester

  • MDL number MFCD07021285
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T16572

Safety Information

MSDS https://www.manchesterorganics.com/T16572
Availability details for
Quantity
  • Availability
  • Add to Cart