MAN182106181 Manchester Organics

5-Pyrazin-2-yl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06738985
  • Molecular Weight : 190.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V16739

Safety Information

MSDS https://www.manchesterorganics.com/V16739
Availability details for
Quantity
  • Availability
  • Add to Cart