MAN182106220 Manchester Organics

4-Fluoro-3-methylbenzonitrile

  • MDL number MFCD03788510
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F16538

Safety Information

MSDS https://www.manchesterorganics.com/F16538
Availability details for
Quantity
  • Availability
  • Add to Cart