MAN182106283 Manchester Organics

5-Bromonicotinoyl chloride

  • MDL number MFCD00173853
  • Molecular Weight : 220.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B16699

Safety Information

MSDS https://www.manchesterorganics.com/B16699
Availability details for
Quantity
  • Availability
  • Add to Cart