MAN182106284 Manchester Organics

3-Propionylpyridine

  • MDL number MFCD00023589
  • Cas Number : 1570-48-5
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-pyridinyl)-1-propanone
Inchi Code 1S/C8H9NO/c1-2-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3
Inchi Key VDNKJMUNLKAGAM-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E16679
Availability details for
Quantity
  • Availability
  • Add to Cart