MAN182106321 Manchester Organics

2-(Pentafluorophenyl)ethanol

  • MDL number MFCD00042331
  • Molecular Weight : 212.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B16816

Safety Information

MSDS https://www.manchesterorganics.com/B16816
Availability details for
Quantity
  • Availability
  • Add to Cart