MAN182106343 Manchester Organics

Benzoylchloride-4-boronic acid

  • MDL number MFCD08062416
  • Molecular Weight : 184.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R16996

Safety Information

MSDS https://www.manchesterorganics.com/R16996
Availability details for
Quantity
  • Availability
  • Add to Cart