MAN182106371 Manchester Organics

6-Bromo-5-nitroisoquinoline

  • MDL number MFCD08282782
  • Molecular Weight : 253.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A17124

Safety Information

MSDS https://www.manchesterorganics.com/A17124
Availability details for
Quantity
  • Availability
  • Add to Cart