MAN182106374 Manchester Organics

4-(Aminomethyl)pyrimidine hydrochloride

  • MDL number MFCD07375466
  • Cas Number : 1138011-17-2
  • Molecular Weight : 145.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J17109
Availability details for
Quantity
  • Availability
  • Add to Cart