MAN182106379 Manchester Organics

4-Pyrimidinemethanol

  • MDL number MFCD03789625
  • Molecular Weight : 110.114

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C17126
Availability details for
Quantity
  • Availability
  • Add to Cart