MAN182106393 Manchester Organics

3-Hydroxyphenylsulfur pentafluoride

  • MDL number MFCD03788515
  • Molecular Weight : 220.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X17167
Availability details for
Quantity
  • Availability
  • Add to Cart