MAN182106453 Manchester Organics

4-Bromo-5-methylthiophene-2-carboxadehyde

  • MDL number MFCD01118659
  • Molecular Weight : 205.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D17292
Availability details for
Quantity
  • Availability
  • Add to Cart