MAN182106485 Manchester Organics

3-(Trifluoromethylthio)aniline

  • MDL number MFCD00190130
  • Molecular Weight : 193.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L17679
Availability details for
Quantity
  • Availability
  • Add to Cart