MAN182106496 Manchester Organics

3-(3-Nitrophenyl)acrylic acid methyl ester

  • MDL number MFCD00024570
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/H17699
Availability details for
Quantity
  • Availability
  • Add to Cart