MAN182106512 Manchester Organics

2-Chloro-5-nitrobenzoyl chloride

  • MDL number MFCD00059180
  • Molecular Weight : 220.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R17896

Safety Information

MSDS https://www.manchesterorganics.com/R17896
Availability details for
Quantity
  • Availability
  • Add to Cart