MAN182106528 Manchester Organics

2,4,6-Trifluoropyridine

  • MDL number MFCD03001161
  • Cas Number : 3512-17-2
  • Molecular Weight : 133.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/S17755
Availability details for
Quantity
  • Availability
  • Add to Cart