MAN182106544 Manchester Organics

4-Chloro-3-fluorophenylacetonitrile

  • MDL number MFCD04115860
  • Molecular Weight : 169.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W17782
Availability details for
Quantity
  • Availability
  • Add to Cart