MAN182106568 Manchester Organics

Methylene bis(toluene-4-sulfonate)

  • MDL number MFCD10568564
  • Cas Number : 24124-59-2
  • Molecular Weight : 356.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/F10787
Availability details for
Quantity
  • Availability
  • Add to Cart