MAN182106569 Manchester Organics

2,5-Bis(trifluoromethyl)benzenesulfonamide

  • MDL number MFCD09475420
  • Molecular Weight : 293.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B10852

Safety Information

MSDS https://www.manchesterorganics.com/B10852
Availability details for
Quantity
  • Availability
  • Add to Cart