MAN182106574 Manchester Organics

1,4-Bis(trimethylsilyl)tetrafluorobenzene

  • MDL number MFCD00093109
  • Cas Number : 16956-91-5
  • Molecular Weight : 294.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin GB
Product Link https://www.manchesterorganics.com/W10540
Availability details for
Quantity
  • Availability
  • Add to Cart