MAN182106580 Manchester Organics

(3-Bromo-4,5-dihydro-isoxazol-5-ylmethyl)carbamic acid tert-butyl ester

  • MDL number MFCD08741480
  • Cas Number : 109770-82-3
  • Molecular Weight : 279.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3-bromo-4,5-dihydro-5-isoxazolyl)methylcarbamate
Inchi Code 1S/C9H15BrN2O3/c1-9(2,3)14-8(13)11-5-6-4-7(10)12-15-6/h4,6,12H,5H2,1-3H3,(H,11,13)
Inchi Key CPTPHEJCUYIFQZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/H10789
Availability details for
Quantity
  • Availability
  • Add to Cart