MAN182106581 Manchester Organics

4-Bromo-2,2-diphenylbutyric acid

  • MDL number MFCD01321300
  • Cas Number : 37742-98-6
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/X10209
Availability details for
Quantity
  • Availability
  • Add to Cart