MAN182106609 Manchester Organics

2-Chloro-6-methylbenzoic acid

  • MDL number MFCD00045799
  • Cas Number : 21327-86-6
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/W10563
Availability details for
Quantity
  • Availability
  • Add to Cart