MAN182106613 Manchester Organics

2-Chloro-5-nitrocinnamic acid

  • MDL number MFCD00063312
  • Cas Number : 36015-19-7
  • Molecular Weight : 227.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-3-(2-chloro-5-nitrophenyl)-2-propenoic acid
Inchi Code 1S/C9H6ClNO4/c10-8-3-2-7(11(14)15)5-6(8)1-4-9(12)13/h1-5H,(H,12,13)/b4-1+
Inchi Key IHTLKLSRRSWIGD-DAFODLJHSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/V10113
Availability details for
Quantity
  • Availability
  • Add to Cart