MAN182106614 Manchester Organics

(4-Chlorophenyl)acetic acid tert-butyl ester

  • MDL number MFCD08687454
  • Cas Number : 33155-59-8
  • Molecular Weight : 226.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/J10694
Availability details for
Quantity
  • Availability
  • Add to Cart