MAN182106619 Manchester Organics

4-Cyanobenzenesulfonamide

  • MDL number MFCD00178767
  • Molecular Weight : 182.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U10728
Availability details for
Quantity
  • Availability
  • Add to Cart