MAN182106638 Manchester Organics

2,2-Difluorocyclopropylamine hydrochloride

  • MDL number MFCD07777150
  • Cas Number : 105614-25-3
  • Molecular Weight : 129.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D10785
Availability details for
Quantity
  • Availability
  • Add to Cart