MAN182106640 Manchester Organics

2,4-Difluorophenylglyoxal hydrate

  • MDL number MFCD04038287
  • Molecular Weight : 188.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J10410
Availability details for
Quantity
  • Availability
  • Add to Cart