MAN182106647 Manchester Organics

3,4-Dimethoxyphenylglyoxal hydrate

  • MDL number MFCD06800598
  • Molecular Weight : 212.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W10872

Safety Information

MSDS https://www.manchesterorganics.com/W10872
Availability details for
Quantity
  • Availability
  • Add to Cart