MAN182106652 Manchester Organics

2,2-Dimethyltetrahydropyran-4-one

  • MDL number MFCD01549337
  • Molecular Weight : 128.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F10762
Availability details for
Quantity
  • Availability
  • Add to Cart