MAN182106665 Manchester Organics

1-(4-Fluorophenyl)-2-methylpropan-1-one

  • MDL number MFCD04038305
  • Cas Number : 26393-91-9
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A10354
Availability details for
Quantity
  • Availability
  • Add to Cart