MAN182106685 Manchester Organics

Methylisonicotinate-N-oxide

  • MDL number MFCD00128856
  • Cas Number : 3783-38-8
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/C10333
Availability details for
Quantity
  • Availability
  • Add to Cart