MAN182106696 Manchester Organics

4-Phenoxybenzenesulfonamide

  • MDL number MFCD00623227
  • Cas Number : 123045-62-5
  • Molecular Weight : 249.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/C10807
Availability details for
Quantity
  • Availability
  • Add to Cart