MAN182106698 Manchester Organics

4-n-Propylpiperidine

  • MDL number MFCD00023151
  • Cas Number : 22398-09-0
  • Molecular Weight : 127.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A10496
Availability details for
Quantity
  • Availability
  • Add to Cart