MAN182106757 Manchester Organics

2-Amino-6-isopropyl-4-pyrimidinol

  • MDL number MFCD02091393
  • Cas Number : 73576-32-6
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-6-isopropyl-4(5H)-pyrimidinone
Inchi Code 1S/C7H11N3O/c1-4(2)5-3-6(11)10-7(8)9-5/h3-4H,1-2H3,(H3,8,9,10,11)
Inchi Key MNIKKPHWWBUBCL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/N10178
Availability details for
Quantity
  • Availability
  • Add to Cart