MAN182106772 Manchester Organics

Diethyl 4-chlorophenyl malonate

  • MDL number MFCD06204602
  • Cas Number : 19677-37-3
  • Molecular Weight : 270.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl 2-(4-chlorophenyl)malonate
Inchi Code 1S/C13H15ClO4/c1-3-17-12(15)11(13(16)18-4-2)9-5-7-10(14)8-6-9/h5-8,11H,3-4H2,1-2H3
Inchi Key PPESOPZGUSEQSO-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/E16986
Availability details for
Quantity
  • Availability
  • Add to Cart